D-Cystine, 98%
D-Cystine, 98%
D-Cystine, 98%
Thermo Scientific Chemicals

D-Cystine, 98%

CAS: 349-46-2 | C6H12N2O4S2 | 240.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L13772.06
also known as L13772-06
Price (USD)
305.65
Special offer
Online exclusive
Ends: 30-Jun-2026
359.00
Save 53.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
305.65
Special offer
Online exclusive
Ends: 30-Jun-2026
359.00
Save 53.35 (15%)
Each
Chemical Identifiers
CAS349-46-2
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Optical Rotation+219° ± 5° (c=1, 1N HCl)
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only