2,4-Dimethoxyphenylacetic acid, 98%
2,4-Dimethoxyphenylacetic acid, 98%
2,4-Dimethoxyphenylacetic acid, 98%
Thermo Scientific Chemicals

2,4-Dimethoxyphenylacetic acid, 98%

CAS: 6496-89-5 | C10H11O4 | 195.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L13784.06
also known as L13784-06
Price (USD)
88.65
Online Exclusive
98.00
Save 9.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
88.65
Online Exclusive
98.00
Save 9.35 (10%)
Each
Chemical Identifiers
CAS6496-89-5
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)107.5-113.5?C
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only