4'-(4-Bromophenyl)acetophenone, 97%
4'-(4-Bromophenyl)acetophenone, 97%
4'-(4-Bromophenyl)acetophenone, 97%
Thermo Scientific Chemicals

4'-(4-Bromophenyl)acetophenone, 97%

CAS: 5731-01-1 | C14H11BrO | 275.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L13798.14
also known as L13798-14
Price (USD)
68.65
Online exclusive
76.10
Save 7.45
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
68.65
Online exclusive
76.10
Save 7.45
Each
Chemical Identifiers
CAS5731-01-1
IUPAC Name1-{4'-bromo-[1,1'-biphenyl]-4-yl}ethan-1-one
Molecular FormulaC14H11BrO
InChI KeyUUVKNCRMWPNBNM-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(C=C1)C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)125.5-134.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only