Quinoline-3-carboxaldehyde, 98+%
Quinoline-3-carboxaldehyde, 98+%
Quinoline-3-carboxaldehyde, 98+%
Thermo Scientific Chemicals

Quinoline-3-carboxaldehyde, 98+%

CAS: 13669-42-6 | C10H7NO | 157.172 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number L13815.06
also known as L13815-06
Price (USD)
391.65
Online Exclusive
435.00
Save 43.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
391.65
Online Exclusive
435.00
Save 43.35 (10%)
Each
Chemical Identifiers
CAS13669-42-6
IUPAC Namequinoline-3-carbaldehyde
Molecular FormulaC10H7NO
InChI KeyRYGIHSLRMNXWCN-UHFFFAOYSA-N
SMILESO=CC1=CN=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Free acid (titration)≤1% (non-U.S. sourced material)
Appearance (Color)White to dark cream to cream to yellow to brown
FormCrystals or powder or crystalline powder or fused solid
CommentMaterial sourced in the U.S. and in other countries
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only