3',4',7,8-Tetramethoxyflavone, 97%
3',4',7,8-Tetramethoxyflavone, 97%
3',4',7,8-Tetramethoxyflavone, 97%
Thermo Scientific Chemicals

3',4',7,8-Tetramethoxyflavone, 97%

CAS: 65548-55-2 | C19H18O6 | 342.347 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
250 mg
Catalog number L13829.MD
also known as L13829-MD
Price (USD)
113.65
Online Exclusive
126.00
Save 12.35 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
113.65
Online Exclusive
126.00
Save 12.35 (10%)
Each
Specifications
Beilstein330821
CAS65548-55-2
Chemical Name or Material3',4',7,8-Tetramethoxyflavone
Formula Weight342.35
IUPAC Name2-(3,4-dimethoxyphenyl)-7,8-dimethoxychromen-4-one
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only