sec-Butyl acetate, 98%
sec-Butyl acetate, 98%
sec-Butyl acetate, 98%
sec-Butyl acetate, 98%
Thermo Scientific Chemicals

sec-Butyl acetate, 98%

CAS: 105-46-4 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L13892.22
also known as L13892-22
Price (USD)
85.60
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
85.60
Each
Add to cart
Chemical Identifiers
CAS105-46-4
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.3870-1.3910 @ 20°C (non-U.S. specification)
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
CommentSpecification differs for U.S. and non-U.S. material where indicated
sec-Butyl acetate is used in organic synthesis, catalytic agent, petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
sec-Butyl acetate is used in organic synthesis, catalytic agent, petrochemical additive.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. P. S. Skell; W. L. Hall. Pyrolysis of sec-Butyl Acetate. Stereospecific cis Elimination. J. Am. Chem. Soc. 1964, 86, (8), 1557-1558.
  2. Wenlin Zhang; Wei Du; Nan Meng; Ruyi Sun; Yongjun Shao; Chunli Li. Isobaric Vapor-liquid Equilibrium of Hexamethyl Disiloxane + Sec-butyl Acetate System at Normal Pressure. Energy Procedia. 2012, 16, 1078-1083.