Bis(2-ethylhexyl)amine, 99%
Bis(2-ethylhexyl)amine, 99%
Bis(2-ethylhexyl)amine, 99%
Bis(2-ethylhexyl)amine, 99%
Thermo Scientific Chemicals

Bis(2-ethylhexyl)amine, 99%

CAS: 106-20-7 | C16H35N | 241.463 g/mol
Have Questions?
Catalog NumberQuantity
L13894.AP
also known as L13894-AP
500 mL
Catalog number L13894.AP
also known as L13894-AP
Price (USD)
88.65
Special offer
Online exclusive
Ends: 30-Jun-2026
104.00
Save 15.35 (15%)
Each
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)
88.65
Special offer
Online exclusive
Ends: 30-Jun-2026
104.00
Save 15.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS106-20-7
IUPAC Namebis(2-ethylhexyl)amine
Molecular FormulaC16H35N
InChI KeySAIKULLUBZKPDA-UHFFFAOYNA-N
SMILESCCCCC(CC)CNCC(CC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4415-1.4445 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Bis(2-ethylhexyl)amine (BEA) was used in the synthesis of stable nanosized CdS particles coated by an oriented monolayer of chemically bonded BEA molecules

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-ethylhexyl)amine (BEA) was used in the synthesis of stable nanosized CdS particles coated by an oriented monolayer of chemically bonded BEA molecules

Solubility
Soluble in water 0.045 g/L @ 20˚C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Pietro Calandra; Angela Ruggirello; Andrea Mele; Vincenzo Turco Liveri; Pietro Calandra; Angela Ruggirello; Andrea Mele; Vincenzo Turco Liveri; Pietro Calandra; Angela Ruggirello; Andrea Mele; Vincenzo Turco Liveri. Self-assembly in surfactant-based liquid mixtures: Bis(2-ethylhexyl)phosphoric acid/bis(2-ethylhexyl)amine systems. Journal of Colloid and Interface Science. Soc. 2010, 348, (1), 183-188.
  2. E. Caponetti; L. Pedone; D. Chillura Martino; V. Pantò; V. Turco Liveri. Materials Science and Engineering: C. Energy Procedia. 2003, 23, (4), 531-539.