Dowex™ 50WX4 100-200 (H)
Dowex™ 50WX4 100-200 (H)
Dowex™ 50WX4 100-200 (H)
Dowex™ 50WX4 100-200 (H)
Thermo Scientific Chemicals

Dowex™ 50WX4 100-200 (H)

CAS: 11113-61-4
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number L13917.18
also known as L13917-18
Price (USD)
55.10
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
55.10
Each
Add to cart
Chemical Identifiers
CAS11113-61-4
MDL NumberMFCD00132725
SpecificationsSpecification SheetSpecification Sheet
Particle Size<88µm: 15% max.
FormPowder or beads
CommentPolystyrene crosslinked with 4% divinylbenzene
Moisture holding capacity64-72%
Appearance (Color)Pale orange to pale brown
View more
Dowex™ 50WX4 100-200 (H) is used for the conversion of D,L-glyceraldehyde 3-phosphate diethyl acetal (barium salt) on treatment with boiling water to free aldehyde D,L-glyceraldehyde 3-phosphate. It is also used to investigate the effect of particle size on drug loading and release profiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dowex|r 50WX4 100-200 (H) is used for the conversion of D,L-glyceraldehyde 3-phosphate diethyl acetal (barium salt) on treatment with boiling water to free aldehyde D,L-glyceraldehyde 3-phosphate. It is also used to investigate the effect of particle size on drug loading and release profiles.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Strongly acidic cation-exchange resin, 4% cross-linked, H+ form, 100-200 mesh (dry).
  2. Elkin, K.; Riviello, J.; Small, H. Improvements in ion reflux: An electrodialytic eluent generation and suppression device for ion chromatography. J. Chromatogr. A 2015, 1403, 63-69.
  3. Tellier, L. E.; Miller, T.; McDevitt, T. C.; Temenoff, J. S. Hydrolysis and sulfation pattern effects on release of bioactive bone morphogenetic protein-2 from heparin-based microparticles. J. Mater. Chem. B 2015, 3 (40), 8001-8009.