(1S)-(+)-Menthyl acetate, 99%
(1S)-(+)-Menthyl acetate, 99%
(1S)-(+)-Menthyl acetate, 99%
Thermo Scientific Chemicals

(1S)-(+)-Menthyl acetate, 99%

CAS: 5157-89-1 | C12H22O2 | 198.306 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L13987.03
also known as L13987-03
Price (USD)
79.65
Online Exclusive
88.80
Save 9.15 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
79.65
Online Exclusive
88.80
Save 9.15 (10%)
Each
Chemical Identifiers
CAS5157-89-1
IUPAC Name5-methyl-2-(propan-2-yl)cyclohexyl acetate
Molecular FormulaC12H22O2
InChI KeyXHXUANMFYXWVNG-UHFFFAOYNA-N
SMILESCC(C)C1CCC(C)CC1OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4450-1.4470 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Optical Rotation+82? ±2? (c=2 in Ethanol)
Assay (GC)≥98.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only