Hydrogen peroxide, 35% w/w aq. soln., stab.
Hydrogen peroxide, 35% w/w aq. soln., stab.
Hydrogen peroxide, 35% w/w aq. soln., stab.
Thermo Scientific Chemicals

Hydrogen peroxide, 35% w/w aq. soln., stab.

CAS: 7722-84-1 | H2O2 | 34.014 g/mol
Have Questions?
Catalog NumberQuantity
L14000.AP
also known as L14000-AP
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L14000.AP
also known as L14000-AP
Price (USD)
52.65
Online Exclusive
58.20
Save 5.55 (10%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
52.65
Online Exclusive
58.20
Save 5.55 (10%)
Each
Chemical Identifiers
CAS7722-84-1
IUPAC Nameperoxol
Molecular FormulaH2O2
InChI KeyMHAJPDPJQMAIIY-UHFFFAOYSA-N
SMILESOO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
CommentStabilized with <1% Proprietary Inhibitor
Assay from Suppliers CofA33.5 - 36.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only