3-(3-Pyridyl)propionic acid, 97%
3-(3-Pyridyl)propionic acid, 97%
3-(3-Pyridyl)propionic acid, 97%
Thermo Scientific Chemicals

3-(3-Pyridyl)propionic acid, 97%

CAS: 3724-19-4 | C8H9NO2 | 151.165 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L14007.06
also known as L14007-06
Price (USD)
65.65
Special offer
Online exclusive
Ends: 30-Jun-2026
76.80
Save 11.15 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
65.65
Special offer
Online exclusive
Ends: 30-Jun-2026
76.80
Save 11.15 (15%)
Each
Chemical Identifiers
CAS3724-19-4
IUPAC Name3-(pyridin-3-yl)propanoic acid
Molecular FormulaC8H9NO2
InChI KeyWDGXIUUWINKTGP-UHFFFAOYSA-N
SMILESOC(=O)CCC1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps or fused solid
Assay (HPLC)≥96.0%
Appearance (Color)White to cream to yellow
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only