Benzyl (S)-(+)-glycidyl ether, 98+%
Benzyl (S)-(+)-glycidyl ether, 98+%
Benzyl (S)-(+)-glycidyl ether, 98+%
Thermo Scientific Chemicals

Benzyl (S)-(+)-glycidyl ether, 98+%

CAS: 16495-13-9 | C10H12O2 | 164.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L14033.03
also known as L14033-03
Price (USD)
37.65
Online Exclusive
42.10
Save 4.45 (11%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
37.65
Online Exclusive
42.10
Save 4.45 (11%)
Each
Chemical Identifiers
CAS16495-13-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.0%
Refractive Index1.5155-1.5195 @ 20?C
Optical Rotation5.2 ± 0.1? ( C=5 in toluene )
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only