1-Aminopiperidine, 97%
1-Aminopiperidine, 97%
1-Aminopiperidine, 97%
Thermo Scientific Chemicals

1-Aminopiperidine, 97%

CAS: 2213-43-6 | C5H12N2 | 100.165 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L14078.22
also known as L14078-22
Price (USD)
543.65
Online Exclusive
604.00
Save 60.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
543.65
Online Exclusive
604.00
Save 60.35 (10%)
Each
Chemical Identifiers
CAS2213-43-6
IUPAC Namepiperidin-1-amine
Molecular FormulaC5H12N2
InChI KeyLWMPFIOTEAXAGV-UHFFFAOYSA-N
SMILESNN1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4735-1.4775 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only