4-Aminobenzamidine dihydrochloride, 97%
4-Aminobenzamidine dihydrochloride, 97%
4-Aminobenzamidine dihydrochloride, 97%
4-Aminobenzamidine dihydrochloride, 97%
Thermo Scientific Chemicals

4-Aminobenzamidine dihydrochloride, 97%

CAS: 2498-50-2 | C7H11Cl2N3 | 208.086 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L14089.06
also known as L14089-06
Price (USD)
99.65
Special offer
Online exclusive
Ends: 31-Dec-2025
117.00
Save 17.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
99.65
Special offer
Online exclusive
Ends: 31-Dec-2025
117.00
Save 17.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS2498-50-2
IUPAC Namedihydrogen 4-aminobenzene-1-carboximidamide dichloride
Molecular FormulaC7H11Cl2N3
InChI KeyGHEHNICLPWTXJC-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].NC(=N)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only