(1R)-(-)-Menthyl acetate, 98%
(1R)-(-)-Menthyl acetate, 98%
(1R)-(-)-Menthyl acetate, 98%
Thermo Scientific Chemicals

(1R)-(-)-Menthyl acetate, 98%

CAS: 2623-23-6 | C12H22O2 | 198.306 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L14095.MD
also known as L14095-MD
Price (USD)
36.10
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
36.10
Each
Chemical Identifiers
CAS2623-23-6
IUPAC Name(1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexyl acetate
Molecular FormulaC12H22O2
InChI KeyXHXUANMFYXWVNG-ADEWGFFLSA-N
SMILESCC(C)[C@@H]1CC[C@@H](C)C[C@H]1OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Optical Rotation-80.5° ± 2.0°
Appearance (Color)Clear, colorless
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only