Amberlite XAD-4 - FAQs
Amberlite XAD-4 - FAQs
View additional product information for Amberlite XAD-4 - FAQs (L14142.36, L14142.22)
2 product FAQs found
What is the structure of the monomer for Amberlite XAD-4 (Cat. No. L14142.22, L14142.36)?
The monomers are styrene and divinylbenzene
Is Amberlite XAD-4 (Cat. No. L14142.22, L14142.36) a styrene-based polymer?
Amberlite XAD-4 is a non-polar divinylbenzene-styrene polymer.