What is the structure of the monomer for Amberlite XAD-4 (Cat. No. L14142.22, L14142.36)?The monomers are styrene and divinylbenzene
Is Amberlite XAD-4 (Cat. No. L14142.22, L14142.36) a styrene-based polymer?Amberlite XAD-4 is a non-polar divinylbenzene-styrene polymer.