4-(Trifluoromethyl)benzhydrol, 97%
4-(Trifluoromethyl)benzhydrol, 97%
4-(Trifluoromethyl)benzhydrol, 97%
Thermo Scientific Chemicals

4-(Trifluoromethyl)benzhydrol, 97%

CAS: 395-23-3 | C14H11F3O | 252.236 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L14147.03
also known as L14147-03
Price (USD)
18.30
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
18.30
Each
Specifications
Beilstein2127184
CAS395-23-3
Chemical Name or Material4-(Trifluoromethyl)benzhydrol
EINECS Number209-917-2
Formula Weight252.23
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only