4',5-Dihydroxy-7-methoxyflavone, 97%
4',5-Dihydroxy-7-methoxyflavone, 97%
4',5-Dihydroxy-7-methoxyflavone, 97%
4',5-Dihydroxy-7-methoxyflavone, 97%
Thermo Scientific Chemicals

4',5-Dihydroxy-7-methoxyflavone, 97%

CAS: 437-64-9 | C16H12O5 | 284.267 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Catalog number L14169.MC
also known as L14169-MC
Price (USD)
159.65
Online Exclusive
177.00
Save 17.35 (10%)
Each
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (USD)
159.65
Online Exclusive
177.00
Save 17.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS437-64-9
IUPAC Name5-hydroxy-2-(4-hydroxyphenyl)-7-methoxy-4H-chromen-4-one
Molecular FormulaC16H12O5
InChI KeyJPMYFOBNRRGFNO-UHFFFAOYSA-N
SMILESCOC1=CC(O)=C2C(=O)C=C(OC2=C1)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormCrystals or powder or crystalline powder
Assay (HPLC)≥96.0%
Melting Point (clear melt)286.0-292.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only