Thermo Scientific Chemicals

4-Dimethylaminobenzoyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: L14176.14
25 g, Each
Thermo Scientific Chemicals

4-Dimethylaminobenzoyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: L14176.14
25 g, Each
Quantity
Catalog number: L14176.14
also known as L14176-14
Price (USD)
Price: 171.00
Online price: 145.65
Your price:
Quantity
-

Chemical Identifiers

CAS
4755-50-4
IUPAC Name
4-(ethylamino)benzoyl chloride
Molecular Formula
C9H10ClNO
InChI Key
XFLQAKBFRUFBJS-UHFFFAOYSA-N
SMILES
CCNC1=CC=C(C=C1)C(Cl)=O
Appearance (Color)
White to cream to yellow to brown
Form
Crystals or powder or crystalline powder or lumps
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%

Description

4-Dimethylaminobenzoyl chloride is used to make other chemicals. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Dimethylaminobenzoyl chloride is used to make other chemicals. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Reacts with water.

Notes
It is sensitive to moisture. Handle and store under an inert atmosphere. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Air and moisture sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text