5,7-Dihydroxyflavone, 98%
5,7-Dihydroxyflavone, 98%
5,7-Dihydroxyflavone, 98%
Thermo Scientific Chemicals

5,7-Dihydroxyflavone, 98%

CAS: 480-40-0 | C15H10O4 | 254.241 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L14178.06
also known as L14178-06
Price (USD)
38.65
Online Exclusive
43.00
Save 4.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
38.65
Online Exclusive
43.00
Save 4.35 (10%)
Each
Chemical Identifiers
CAS480-40-0
IUPAC Name5,7-dihydroxy-2-phenyl-4H-chromen-4-one
Molecular FormulaC15H10O4
InChI KeyRTIXKCRFFJGDFG-UHFFFAOYSA-N
SMILESOC1=CC(O)=C2C(=O)C=C(OC2=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
Appearance (Color)Yellow to brown
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only