5-Hydroxyflavone, 97%
5-Hydroxyflavone, 97%
5-Hydroxyflavone, 97%
5-Hydroxyflavone, 97%
Thermo Scientific Chemicals

5-Hydroxyflavone, 97%

CAS: 491-78-1 | C15H10O3 | 238.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Catalog number L14187.ME
also known as L14187-ME
Price (USD)
126.65
Online Exclusive
140.00
Save 13.35 (10%)
Each
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)
126.65
Online Exclusive
140.00
Save 13.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS491-78-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormPowder
Assay (HPLC)≥96.0%
Melting Point (clear melt)154.0-163.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only