(R)-(-)-Epichlorohydrin, 98+%
(R)-(-)-Epichlorohydrin, 98+%
(R)-(-)-Epichlorohydrin, 98+%
Thermo Scientific Chemicals

(R)-(-)-Epichlorohydrin, 98+%

CAS: 51594-55-9 | C3H5ClO | 92.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L14198.06
also known as L14198-06
Price (USD)
134.65
Online Exclusive
149.00
Save 14.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
134.65
Online Exclusive
149.00
Save 14.35 (10%)
Each
Chemical Identifiers
CAS51594-55-9
IUPAC Name(2R)-2-(chloromethyl)oxirane
Molecular FormulaC3H5ClO
InChI KeyBRLQWZUYTZBJKN-VKHMYHEASA-N
SMILESClC[C@H]1CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.0% (non-U.S. specification)
CommentPurchased in the U.S. and in other countries
FormLiquid
Assay from Supplier's CofA≥98.0% (GC, U.S. Specification)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only