N-Ethylcyclohexylamine, 97%
N-Ethylcyclohexylamine, 97%
N-Ethylcyclohexylamine, 97%
Thermo Scientific Chemicals

N-Ethylcyclohexylamine, 97%

CAS: 5459-93-8 | C8H17N | 127.231 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number L14221.AE
also known as L14221-AE
Price (USD)
41.90
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
41.90
Each
Chemical Identifiers
CAS5459-93-8
IUPAC NameN-ethylcyclohexanamine
Molecular FormulaC8H17N
InChI KeyAGVKXDPPPSLISR-UHFFFAOYSA-N
SMILESCCNC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
FormLiquid
Refractive Index1.4500-1.4550 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only