Diethoxymethylvinylsilane, 97%
Diethoxymethylvinylsilane, 97%
Diethoxymethylvinylsilane, 97%
Thermo Scientific Chemicals

Diethoxymethylvinylsilane, 97%

CAS: 5507-44-8 | C7H16O2Si | 160.288 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L14224.22
also known as L14224-22
Price (USD)
123.65
Special offer
Online exclusive
Ends: 30-Jun-2026
145.00
Save 21.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
123.65
Special offer
Online exclusive
Ends: 30-Jun-2026
145.00
Save 21.35 (15%)
Each
Chemical Identifiers
CAS5507-44-8
IUPAC Nameethenyldiethoxymethylsilane
Molecular FormulaC7H16O2Si
InChI KeyMBGQQKKTDDNCSG-UHFFFAOYSA-N
SMILESCCO[Si](C)(OCC)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Infrared spectrumConforms
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only