1,2-Diethoxyethane, 98%
1,2-Diethoxyethane, 98%
1,2-Diethoxyethane, 98%
Thermo Scientific Chemicals

1,2-Diethoxyethane, 98%

CAS: 629-14-1 | C6H14O2 | 118.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L14283.AE
also known as L14283-AE
Price (USD)
-
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS629-14-1
IUPAC Name1,2-diethoxyethane
Molecular FormulaC6H14O2
InChI KeyLZDKZFUFMNSQCJ-UHFFFAOYSA-N
SMILESCCOCCOCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥97.5%
Free acid (titration)(as acetic acid) <0.15%
Water Content (Karl Fischer Titration)<1.0%
Refractive Index1.3900-1.3940 @ 20?C
View more
Ethylene glycol diethyl ether is a important solvent, used for ink, paint and coating industry. Mainly used in polymer, electrochemistry, boracium chemistry, resin, nitro cellulose, In this product , the benzene, xenene, terphenyl, anthracene reacts with sodium and then bring about the complex; it is also used in surface treatment, halogen analysis in gasoline, acetic acid recycle in diluted acetic acid, depaint agent, thinner, flushing agent, stabilizer, anti oxidant , thickener of lube.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylene glycol diethyl ether is a important solvent, used for ink, paint and coating industry. Mainly used in polymer, electrochemistry, boracium chemistry, resin, nitro cellulose, In this product , the benzene, xenene, terphenyl, anthracene reacts with sodium and then bring about the complex; it is also used in surface treatment, halogen analysis in gasoline, acetic acid recycle in diluted acetic acid, depaint agent, thinner, flushing agent, stabilizer, anti oxidant , thickener of lube.

Solubility
Soluble in water 34 g/L @ 20˚C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Amitava Sarkar; Robert A. Keogh; Shiqi Bao; Burtron H. Davis. Fischer-Tropsch synthesis with promoted iron catalyst: Reaction pathways for acetic acid, glycol, 2-ethoxyethanol and 1,2-diethoxyethane. Applied Catalysis A: General. 1979, 341, (1-2), 146-153.
  2. K. Takeuchi; M. Salmeron; G.A. Somorjai.The bonding of hydrogenated and fluorinated diethers to Pt(111) and to zirconium oxide thin film. Surface Science. 1992, 279, (3), 328-340.