Methacrylic anhydride, 94%, stab. with ca 0.2% 2,4-dimethyl-6-tert-butylphenol
Methacrylic anhydride, 94%, stab. with ca 0.2% 2,4-dimethyl-6-tert-butylphenol
Methacrylic anhydride, 94%, stab. with ca 0.2% 2,4-dimethyl-6-tert-butylphenol
Methacrylic anhydride, 94%, stab. with ca 0.2% 2,4-dimethyl-6-tert-butylphenol
Thermo Scientific Chemicals

Methacrylic anhydride, 94%, stab. with ca 0.2% 2,4-dimethyl-6-tert-butylphenol

CAS: 760-93-0 | C8H10O3 | 154.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number L14357.30
also known as L14357-30
Price (USD)
99.10
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
99.10
Each
Add to cart
Chemical Identifiers
CAS760-93-0
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Refractive Index1.4525-1.4605 @ 20°C
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥92.5%
Methacrylic anhydride is a reactive monomer used for the preparation of different type of monomers under mild reaction conditions. It finds applications in light-curing coatings, cross-linked materials like synthetic resin. It acts as an intermediate for the preparation of fibers and resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methacrylic anhydride is a reactive monomer used for the preparation of different type of monomers under mild reaction conditions. It finds applications in light-curing coatings, cross-linked materials like synthetic resin. It acts as an intermediate for the preparation of fibers and resins.

Solubility
Miscible with ethanol and ether.

Notes
Moisture and light sensitive. Store in a cool place. Incompatible with acids, strong oxidizing agents, alcohols, amines and bases.
RUO – Research Use Only

General References:

  1. Erwin, Z.; Blokzijl, M. M.; Grijpma, D. W. A Combinatorial Photocrosslinking Method for the Preparation of Porous Structures with Widely Differing Properties. Macromol. Rapid Commun. 2015, 36 (21), 1902-1909.
  2. Li, M.; Hu, Y.; Li, Y.; Liu, Y.; Zeng, C.; Liu, Q.; Hou, C. Synthesis and Characterization of Polymerizable Tourmaline Methacrylate. e-J. Surf. Sci. Nanotechnol. 2015, 13, 422-425.