3',5-Dihydroxy-4',6,7-trimethoxyflavone, 97%
3',5-Dihydroxy-4',6,7-trimethoxyflavone, 97%
3',5-Dihydroxy-4',6,7-trimethoxyflavone, 97%
Thermo Scientific Chemicals

3',5-Dihydroxy-4',6,7-trimethoxyflavone, 97%

CAS: 855-96-9 | C18H16O7 | 344.319 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Catalog number L14495.MB
also known as L14495-MB
Price (USD)
270.65
Special offer
Online exclusive
Ends: 28-Dec-2026
318.00
Save 47.35
Each
Quantity:
25 mg
Request bulk or custom format
Price (USD)
270.65
Special offer
Online exclusive
Ends: 28-Dec-2026
318.00
Save 47.35
Each
Chemical Identifiers
CAS855-96-9
IUPAC Name5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-6,7-dimethoxy-4H-chromen-4-one
Molecular FormulaC18H16O7
InChI KeyKLAOKWJLUQKWIF-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1O)C1=CC(=O)C2=C(O)C(OC)=C(OC)C=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)188.0-197.0?C
Appearance (Color)White to yellow to brown
FormPowder
Assay (HPLC)≥96.0%
3',5-Dihydroxy-4',6,7-trimethoxyflavone is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′,5-Dihydroxy-4′,6,7-trimethoxyflavone is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water. Soluble in chloroform and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with and oxidizing agents.
RUO – Research Use Only

General References:

  1. G.A Akowuah, , I Zhari, I Norhayati, A Sadikun, S.M Khamsah. Sinensetin, eupatorin, 3'-hydroxy-5, 6, 7, 4'-tetramethoxyflavone and rosmarinic acid contents and antioxidative effect of Orthosiphon stamineus from Malaysia. Food Chemistry. 2004, 87 (4), 559-566.
  2. Vasilis P. Androutsopoulo.;Naichang Li.; Randolph R. J. Arroo. The Methoxylated Flavones Eupatorin and Cirsiliol Induce CYP1 Enzyme Expression in MCF7 Cells. J. Nat. Prod. 2009, 72 (8), 1390-1394.