N,N-Dimethylcyclohexylamine, 98+%
N,N-Dimethylcyclohexylamine, 98+%
N,N-Dimethylcyclohexylamine, 98+%
Thermo Scientific Chemicals

N,N-Dimethylcyclohexylamine, 98+%

CAS: 98-94-2 | C8H17N | 127.231 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number L14521.AC
also known as L14521-AC
Price (USD)
28.65
Online exclusive
31.80
Save 3.15
Each
Quantity:
25 mL
Request bulk or custom format
Price (USD)
28.65
Online exclusive
31.80
Save 3.15
Each
Chemical Identifiers
CAS98-94-2
IUPAC NameN,N-dimethylcyclohexanamine
Molecular FormulaC8H17N
InChI KeySVYKKECYCPFKGB-UHFFFAOYSA-N
SMILESCN(C)C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4520-1.4560 @ 20?C
Assay (GC)≥98.0%
FormLiquid
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only