Filter Aid, Celite Standard Supercel∣r
Filter Aid, Celite Standard Supercel∣r
Filter Aid, Celite Standard Supercel∣r
Thermo Scientific Chemicals

Filter Aid, Celite Standard Supercel∣r

CAS: 61790-53-2 | O2Si | 60.083 g/mol
Have Questions?
Catalog NumberQuantity
L14552.36
also known as L14552-36
500 g
Catalog number L14552.36
also known as L14552-36
Price (USD)
72.65
Online Exclusive
80.30
Save 7.65 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
72.65
Online Exclusive
80.30
Save 7.65 (10%)
Each
Chemical Identifiers
CAS61790-53-2
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Particle Size+140 mesh ASTM (100?m, 150mesh Tyler)
CommentCentrifuged Wet Density : ca 310 g/L
Appearance (Color)White to pale brown or pink
CommentPermeability : ca 0.25 Darcys

Filter Aid, Celite AW Standard Supercel™ is used for general filtrations. It is also used as a functional filler in paints, plastics, paper, rubber, adhesives, catalysts, agricultural chemicals, pharmaceuticals and other chemicals, and a mild abrasive in polishes and cleaners. It is also used in clarifying liquids and oils, as an efficient thermal insulator; in manufacture of heat insulators, fire brick, and fire- and acid-proof packing materials; as an absorbent; in absorbent dynamite; as a solid carrier in chromatography; pozzolanic admixture in concrete mixes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Filter Aid, Celite AW Standard Supercel« is used for general filtrations. It is also used as a functional filler in paints, plastics, paper, rubber, adhesives, catalysts, agricultural chemicals, pharmaceuticals and other chemicals,
RUO – Research Use Only

General References:

  1. S. E Calvert. Accumulation of Diatomaceous Silica in the Sediments of the Gulf of California. Geological Society of America Bulletin. 1965, 77 (6), 569-596.
  2. Engel G. Vrieling; Theo P. M. Beelen; Rutger A. Van Santen andWinfried W. C. Gieskes. Nanoscale uniformity of pore architecture in diatomaceous silica: a combined small and wide angle x-ray scattering study. Journal of Phycology. 2000, 36 (1), 146-159.
  3. Celite is a registered trademark of Celite Corp.
  4. Acid-washed grade for general filtrations.