2-Amino-6-methyl-4(3H)-quinazolone, 96%, Thermo Scientific Chemicals
2-Amino-6-methyl-4(3H)-quinazolone, 96%, Thermo Scientific Chemicals
2-Amino-6-methyl-4(3H)-quinazolone, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-6-methyl-4(3H)-quinazolone, 96%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
L14558.03
also known as L14558-03
1 g
Catalog number L14558.03
also known as L14558-03
Price (USD)
123.65
Online Exclusive
137.00
Save 13.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
123.65
Online Exclusive
137.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS50440-82-9
IUPAC Name2-amino-6-methyl-1,4-dihydroquinazolin-4-one
Molecular FormulaC9H9N3O
InChI KeyDJYDUJAKTAWKTN-UHFFFAOYSA-N
SMILESCC1=CC=C2NC(N)=NC(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
FormPowder
Assay (HPLC)≥95.0%
It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
Dennis J. McNamara; Ellen M. Berman; David W. Fry; Leslie M. Werbel. Potent inhibition of thymidylate synthase by two series of nonclassical quinazolines. J. Med. Chem.1990, 33(7), 2045-2051.