(R)-2-Methyl-CBS-oxazaborolidine, 1M soln. in toluene
(R)-2-Methyl-CBS-oxazaborolidine, 1M soln. in toluene
(R)-2-Methyl-CBS-oxazaborolidine, 1M soln. in toluene
Thermo Scientific Chemicals

(R)-2-Methyl-CBS-oxazaborolidine, 1M soln. in toluene

CAS: 112022-83-0 | C18H20BNO | 277.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L14582.AA
also known as L14582-AA
Price (USD)
82.65
Online Exclusive
91.90
Save 9.25 (10%)
Each
Quantity:
5 mL
Request bulk or custom format
Price (USD)
82.65
Online Exclusive
91.90
Save 9.25 (10%)
Each
Chemical Identifiers
CAS112022-83-0
IUPAC Name(3aR)-1-methyl-3,3-diphenyl-hexahydropyrrolo[1,2-c][1,3,2]oxazaborole
Molecular FormulaC18H20BNO
InChI KeyVMKAFJQFKBASMU-QGZVFWFLSA-N
SMILES[H][C@]12CCCN1B(C)OC2(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange
FormLiquid
Assay (Non-aqueous acid-base Titration)0.90-1.10M
(R)-2-Methyl-CBS-oxazaborolidine is used in the asymmetric reduction of prochiral ketones. Other applications include the enantioselective synthesis of α-hydroxy acids, α-amino acids, C2 symmetrical ferrocenyl diols, and propargyl alcohols. It is also used in a desymmetrizing reduction leading to (S)-4-hydroxycyclohexenone. It is useful in the production of stereospecific motifs such as α-hydroxy acids, α-amino acids, symmetrical ferrocenyl diols and propargyl alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-2-Methyl-CBS-oxazaborolidine is used in the asymmetric reduction of prochiral ketones. Other applications include the enantioselective synthesis of α-hydroxy acids, α-amino acids, C2 symmetrical ferrocenyl diols, and propargyl alcohols. It is also used in a desymmetrizing reduction leading to (S)-4-hydroxycyclohexenone. It is useful in the production of stereospecific motifs such as α-hydroxy acids, α-amino acids, symmetrical ferrocenyl diols and propargyl alcohols.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store under inert gas. It is sensitive to moisture and air. Incompatible with oxidizing agents.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Parker, K. A.; Ledeboer, M. W. Asymmetric Reduction. A Convenient Method for the Reduction of Alkynyl Ketones. J. Org. Chem. 1996, 61 3214.
  2. Marcel Hoogenraad.; Guido M Klaus.; Niels Elders.; Simone M Hooijschuur.; Ben McKay.; Alan A Smith.; Eric W.P Damen. Oxazaborolidine mediated asymmetric ketone reduction: prediction of enantiomeric excess based on catalyst structure. Tetrahedron: Asymmetry. 2004, 15 (3), 519-523.
  3. Anhydrous catalyst for CBS reduction reaction. For further information, see (S)-2-Methyl-CBS-oxazaborolidine monohydrate, L09219.