Valeramide, 97%
Valeramide, 97%
Valeramide, 97%
Thermo Scientific Chemicals

Valeramide, 97%

CAS: 626-97-1 | C5H11NO | 101.149 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L14601.06
also known as L14601-06
Price (USD)
39.80
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
39.80
Each
Chemical Identifiers
CAS626-97-1
IUPAC Namepentanamide
Molecular FormulaC5H11NO
InChI KeyIPWFJLQDVFKJDU-UHFFFAOYSA-N
SMILESCCCCC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder or flakes
Melting Point (clear melt)100.5-106.5?C
Valeramide is used in r GC (Gas Chromatography) and LC (Liquid Chromatography) analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Valeramide is used in r GC (Gas Chromatography) and LC (Liquid Chromatography) analysis.

Solubility
Soluble in water (almost transparency).

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Jessica Loos; Detlef Schröder; Waltraud Zummack; Helmut Schwarz. Regio- and diastereoselective C-H bond activation of valeramide and 3-methyl valeramide by bare Fe+ ions. International Journal of Mass Spectrometry.2002, 217(1-3), 169-177.
  2. W. E. Dulin; F. L. Schmidt; M. C. Blanks; G. H. Lund. Hypoglycemic Activity of Valeramide, 4-Dimethylamino-N-methyl-2, 2-Diphenyl-, Hydrochloride. Exp Biol Med (Maywood).1962, 109(3), 721-724.