6-Methoxy-2-naphthoic acid, 98+%
6-Methoxy-2-naphthoic acid, 98+%
6-Methoxy-2-naphthoic acid, 98+%
Thermo Scientific Chemicals

6-Methoxy-2-naphthoic acid, 98+%

CAS: 2471-70-7 | C12H9O3 | 201.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L14610.03
also known as L14610-03
Price (USD)
34.65
Special offer
Online exclusive
Ends: 30-Jun-2026
41.00
Save 6.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
34.65
Special offer
Online exclusive
Ends: 30-Jun-2026
41.00
Save 6.35 (15%)
Each
Chemical Identifiers
CAS2471-70-7
IUPAC Name6-methoxynaphthalene-2-carboxylate
Molecular FormulaC12H9O3
InChI KeyYZBILXXOZFORFE-UHFFFAOYSA-M
SMILESCOC1=CC=C2C=C(C=CC2=C1)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Melting Point198-205?C
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only