Quinoline-4-carboxylic acid, 98+%
Quinoline-4-carboxylic acid, 98+%
Quinoline-4-carboxylic acid, 98+%
Thermo Scientific Chemicals

Quinoline-4-carboxylic acid, 98+%

CAS: 486-74-8 | C10H7NO2 | 173.171 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L14611.06
also known as L14611-06
Price (USD)
186.65
Special offer
Online exclusive
Ends: 30-Jun-2026
219.00
Save 32.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
186.65
Special offer
Online exclusive
Ends: 30-Jun-2026
219.00
Save 32.35 (15%)
Each
Specifications
Beilstein5224
CAS486-74-8
ChEBICHEBI:18311
Chemical Name or MaterialQuinoline-4-carboxylic acid
EINECS Number207-640-1
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only