Cycloheptanemethylamine, 98+%
Cycloheptanemethylamine, 98+%
Cycloheptanemethylamine, 98+%
Cycloheptanemethylamine, 98+%
Thermo Scientific Chemicals

Cycloheptanemethylamine, 98+%

CAS: 4448-77-5 | C8H17N | 127.231 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L14659.03
also known as L14659-03
Price (USD)
51.65
Online Exclusive
57.60
Save 5.95 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
51.65
Online Exclusive
57.60
Save 5.95 (10%)
Each
Add to cart
Chemical Identifiers
CAS4448-77-5
IUPAC Name1-cycloheptylmethanamine
Molecular FormulaC8H17N
InChI KeyCAOQEOHEZKVYOJ-UHFFFAOYSA-N
SMILESNCC1CCCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Water Content (Karl Fischer Titration)<1.0%
Assay (GC)>98.0%
Refractive Index1.4715-1.4765 @ 20?C
It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible or difficult to mix with water.

Notes
Air Sensitive, Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only