2,5-Bis(trifluoromethyl)aniline, 99%
2,5-Bis(trifluoromethyl)aniline, 99%
2,5-Bis(trifluoromethyl)aniline, 99%
Thermo Scientific Chemicals

2,5-Bis(trifluoromethyl)aniline, 99%

CAS: 328-93-8 | C8H5F6N | 229.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L14706.06
also known as L14706-06
Price (USD)
112.65
Online Exclusive
125.00
Save 12.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
112.65
Online Exclusive
125.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS328-93-8
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4320-1.4350 @ 20°C
Assay (GC)≥98.5%
Appearance (Color)Clear colorless to yellow to orange
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only