2-Chloro-6-fluorobenzaldoxime, 97%
2-Chloro-6-fluorobenzaldoxime, 97%
2-Chloro-6-fluorobenzaldoxime, 97%
Thermo Scientific Chemicals

2-Chloro-6-fluorobenzaldoxime, 97%

CAS: 443-33-4 | C7H5ClFNO | 173.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L14719.06
also known as L14719-06
Price (USD)
62.90
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
62.90
Each
Specifications
Beilstein3247886
CAS443-33-4
Chemical Name or Material2-Chloro-6-fluorobenzaldoxime
EINECS Number207-135-6
Formula Weight173.58
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only