3,5-Dimethylbenzyl alcohol, 99%
3,5-Dimethylbenzyl alcohol, 99%
3,5-Dimethylbenzyl alcohol, 99%
3,5-Dimethylbenzyl alcohol, 99%
Thermo Scientific Chemicals

3,5-Dimethylbenzyl alcohol, 99%

CAS: 27129-87-9 | C9H12O | 136.194 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L14726.03
also known as L14726-03
Price (USD)
35.65
Online Exclusive
39.20
Save 3.55 (9%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
35.65
Online Exclusive
39.20
Save 3.55 (9%)
Each
Add to cart
Chemical Identifiers
CAS27129-87-9
IUPAC Name(3,5-dimethylphenyl)methanol
Molecular FormulaC9H12O
InChI KeyIQWWTJDRVBWBEL-UHFFFAOYSA-N
SMILESCC1=CC(CO)=CC(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5295-1.5325 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
FormLiquid or viscous liquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only