4-Trimethylsilyl-3-butyn-2-ol, 97%
4-Trimethylsilyl-3-butyn-2-ol, 97%
4-Trimethylsilyl-3-butyn-2-ol, 97%
Thermo Scientific Chemicals

4-Trimethylsilyl-3-butyn-2-ol, 97%

CAS: 6999-19-5 | C7H14OSi | 142.273 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L14815.14
also known as L14815-14
Price (USD)
215.65
Online Exclusive
239.00
Save 23.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
215.65
Online Exclusive
239.00
Save 23.35 (10%)
Each
Specifications
Beilstein1923632
Boiling Point126°C to 129°C
CAS6999-19-5
Chemical Name or Material4-Trimethylsilyl-3-butyn-2-ol
DOT InformationTransport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only