1-Methyl-1H-pyrazole, 97+%
1-Methyl-1H-pyrazole, 97+%
1-Methyl-1H-pyrazole, 97+%
Thermo Scientific Chemicals

1-Methyl-1H-pyrazole, 97+%

CAS: 930-36-9 | C4H6N2 | 82.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L14831.14
also known as L14831-14
Price (USD)
164.65
Online Exclusive
183.00
Save 18.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
164.65
Online Exclusive
183.00
Save 18.35 (10%)
Each
Chemical Identifiers
CAS930-36-9
IUPAC Name1-methyl-1H-pyrazole
Molecular FormulaC4H6N2
InChI KeyUQFQONCQIQEYPJ-UHFFFAOYSA-N
SMILESCN1C=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4730-1.4780 @ 20?C
FormLiquid
Assay (GC)>97.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only