Fumaronitrile, 98+%
Fumaronitrile, 98+%
Fumaronitrile, 98+%
Fumaronitrile, 98+%
Thermo Scientific Chemicals

Fumaronitrile, 98+%

CAS: 764-42-1 | C4H2N2 | 78.074 g/mol
Have Questions?
Catalog NumberQuantity
L14911.06
also known as L14911-06
5 g
Catalog number L14911.06
also known as L14911-06
Price (USD)
68.65
Special offer
Online exclusive
Ends: 30-Jun-2026
80.60
Save 11.95 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
68.65
Special offer
Online exclusive
Ends: 30-Jun-2026
80.60
Save 11.95 (15%)
Each
Chemical Identifiers
CAS764-42-1
IUPAC Name(2E)-but-2-enedinitrile
Molecular FormulaC4H2N2
InChI KeyKYPOHTVBFVELTG-OWOJBTEDSA-N
SMILESN#C\C=C\C#N
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Identification (FTIR)Conforms
Appearance (Color)White to pale cream
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only