Methacrylamide, 98%
Methacrylamide, 98%
Methacrylamide, 98%
Thermo Scientific Chemicals

Methacrylamide, 98%

CAS: 79-39-0 | C4H7NO | 85.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number L15013.22
also known as L15013-22
Price (USD)
31.10
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
31.10
Each
Chemical Identifiers
CAS79-39-0
IUPAC Name2-methylprop-2-enamide
Molecular FormulaC4H7NO
InChI KeyFQPSGWSUVKBHSU-UHFFFAOYSA-N
SMILESCC(=C)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystalline powder or crystals
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)107-117?C
Methacrylamide is a reagent used in the production of polymers and copolymers. It is also used as a raw material in the production of chemicals for industrial and professional use such as textiles, leather, fur, bulk, large scale chemicals (including petroleum products), fine chemicals, formulation [mixing] of preparations and/or re-packaging (excluding alloys), building and construction work, electricity, steam, gas, water supply and sewage treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methacrylamide is a reagent used in the production of polymers and copolymers. It is also used as a raw material in the production of chemicals for industrial and professional use such as textiles, leather, fur, bulk, large scale chemicals (including petroleum products), fine chemicals, formulation [mixing] of preparations and/or re-packaging (excluding alloys), building and construction work, electricity, steam, gas, water supply and sewage treatment.

Solubility
Soluble in water (202g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Laura M. Y. Yu.; Karineh Kazazian.; Molly S. Shoichet. Peptide surface modification of methacrylamide chitosan for neural tissue engineering applications. Journal of Biomedical Materials Research Part A. 2007, 82A (1), 243-255.
  2. Burcin Akgun.; Emre Savci.; Duygu Avci. Synthesis and polymerizations of phosphonated-bis(methacrylamide)s for dental applications. Journal of Polymer Science Part A: Polymer Chemistry. 2012, 50 (4), 801-810.