1-Methyl-1-cyclohexene, 96%
1-Methyl-1-cyclohexene, 96%
1-Methyl-1-cyclohexene, 96%
Thermo Scientific Chemicals

1-Methyl-1-cyclohexene, 96%

CAS: 591-49-1 | C7H12 | 96.173 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L15014.14
also known as L15014-14
Price (USD)
96.65
Special offer
Online exclusive
Ends: 30-Jun-2026
114.00
Save 17.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
96.65
Special offer
Online exclusive
Ends: 30-Jun-2026
114.00
Save 17.35 (15%)
Each
Chemical Identifiers
CAS591-49-1
IUPAC Name1-methylcyclohex-1-ene
Molecular FormulaC7H12
InChI KeyCTMHWPIWNRWQEG-UHFFFAOYSA-N
SMILESCC1=CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
Assay (GC)>95.0%
CommentMaterial sourced in the U.S. and in other countries
Refractive Index1.4420-1.4520 @ 20°C (non-U.S. sourced material)
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only