4',5,7-Trihydroxyflavone, 97%
4',5,7-Trihydroxyflavone, 97%
4',5,7-Trihydroxyflavone, 97%
Thermo Scientific Chemicals

4',5,7-Trihydroxyflavone, 97%

CAS: 520-36-5 | C15H10O5 | 270.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Catalog number L15041.MB
also known as L15041-MB
Price (USD)
53.65
Special offer
Online exclusive
Ends: 28-Dec-2026
63.30
Save 9.65
Each
Quantity:
25 mg
Request bulk or custom format
Price (USD)
53.65
Special offer
Online exclusive
Ends: 28-Dec-2026
63.30
Save 9.65
Each
Chemical Identifiers
CAS520-36-5
IUPAC Name5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one
Molecular FormulaC15H10O5
InChI KeyKZNIFHPLKGYRTM-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C1=CC(=O)C2=C(O)C=C(O)C=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to yellow to brown
Assay (HPLC)≥96.0%
FormPowder
Melting Point (clear melt)295.0-305.0?C
4',5,7-Trihydroxyflavone is a nonmutagenic flavonoid shown to inhibit cell proliferation, angiogenesis and protein kinase. Also induces apoptosis in breast cancer cells. It inhibit topoisomerase I-catalyzed DNA re-ligation and enhance gap junctional intercellular communication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′,5,7-Trihydroxyflavone is a nonmutagenic flavonoid shown to inhibit cell proliferation, angiogenesis and protein kinase. Also induces apoptosis in breast cancer cells. It inhibit topoisomerase I-catalyzed DNA re-ligation and enhance gap junctional intercellular communication.

Solubility
Soluble in dimethyl sulfoxide (27 mg/ml) or 1M potassium hydroxide (50mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Hirano T.; Oka K.; Akiba M. Antiproliferative effects of synthetic and naturally occurring flavonoids on tumor cells of the human breast carcinoma cell line, ZR-75-1. Research Communications in Chemical Pathology and Pharmacology. 1989, 64 (1), 69-78.
  2. Sunanda Panda.; Anand Kar. Apigenin (4‘,5,7-trihydroxyflavone) regulates hyperglycaemia, thyroid dysfunction and lipid peroxidation in alloxan-induced diabetic mice. Journal of Pharmacy and Pharmacology. 2007, 59 (11), 1543-1548.