Triethylborane, 1M soln. in THF
Triethylborane, 1M soln. in THF
Triethylborane, 1M soln. in THF
Thermo Scientific Chemicals

Triethylborane, 1M soln. in THF

CAS: 97-94-9 | C6H15B | 98.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number L15109.AC
also known as L15109-AC
Price (USD)
55.80
Each
Quantity:
25 mL
Request bulk or custom format
Price (USD)
55.80
Each
Chemical Identifiers
CAS97-94-9
IUPAC Nametriethylborane
Molecular FormulaC6H15B
InChI KeyLALRXNPLTWZJIJ-UHFFFAOYSA-N
SMILESCCB(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA0.95-1.30 M
Appearance (Color)Clear colorless to yellow to orange
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Triethylborane is catalyst for allylation of aldehydes, decarboxylative C-C bond cleavage reactions, rhenium hydride / boron Lewis acid cocatalysis of alkene hydrogenations, Regioselective hydroxyalkylation of unsaturated oxime ethers. Reactant for radical reductions of alkyl bromides with N-heterocyclic carbene boranes and synthesis of tetramethylammonium trialkylphenylborate salts with oxidation potential.

Solubility
Reacts with water.

Notes
Air Sensitive, store away form air. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Yanfeng Jiang; Jeannine Hess; Thomas Fox; Heinz Berke. Rhenium hydride/boron Lewis acid cocatalysis of alkene hydrogenations: activities comparable to those of precious metal systems. Journal of the American Chemical Society.2010, 132,(51), 18233-18247.
  2. Masafumi Ueda et. al. Aerobic hydroxylation of N-borylenamine: triethylborane-mediated hydroxyalkylation of alpha,beta-unsaturated oxime ether. Organic Letters,2009, 11(20), 4632-4635.
  3. Radical initiator for hydrostannylation of alkynes: Tetrahedron, 45, 923 (1989).
  4. Reacts with metal enolates to give the enoxytriethylborates, useful in selective alkylation and aldol reactions: Tetrahedron Lett., 2975 (1976); 32, 5521 (1991).