(-)-Fenchone, 98+%
(-)-Fenchone, 98+%
(-)-Fenchone, 98+%
Thermo Scientific Chemicals

(-)-Fenchone, 98+%

CAS: 7787-20-4 | C10H16O | 152.237 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number L15151.18
also known as L15151-18
Price (USD)
42.80
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
42.80
Each
Chemical Identifiers
CAS7787-20-4
IUPAC Name(1R,4S)-1,3,3-trimethylbicyclo[2.2.1]heptan-2-one
Molecular FormulaC10H16O
InChI KeyLHXDLQBQYFFVNW-OIBJUYFYSA-N
SMILESCC1(C)[C@H]2CC[C@](C)(C2)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Optical Rotation-47.5° to -53.5° (neat)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only