What is the bulk density of Montmorillonite K10 (Cat. Nos. L15160.30, L15160-30)?The bulk density of Montmorillonite K10 is 300-370 g/l.
What is the loss of ignition (%) of Montmorillonite K10 (Cat. Nos. L15160.30, L15160-30)?The loss of ignition of Montmorillonite K10 is 7%.