3,4,5-Trimethoxybenzeneboronic acid, 98%
3,4,5-Trimethoxybenzeneboronic acid, 98%
3,4,5-Trimethoxybenzeneboronic acid, 98%
Thermo Scientific Chemicals

3,4,5-Trimethoxybenzeneboronic acid, 98%

CAS: 182163-96-8 | C9H13BO5 | 212.008 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L15191.03
also known as L15191-03
Price (USD)
57.65
Online exclusive
63.80
Save 6.15
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
57.65
Online exclusive
63.80
Save 6.15
Each
Chemical Identifiers
CAS182163-96-8
IUPAC Name(3,4,5-trimethoxyphenyl)boronic acid
Molecular FormulaC9H13BO5
InChI KeyRULQUTYJXDLRFL-UHFFFAOYSA-N
SMILESCOC1=CC(=CC(OC)=C1OC)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown
FormPowder or fused solid or granules
Assay (Aqueous acid-base Titration)≥97.5%
Proton NMRConforms to structure
Identification (FTIR)Conforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only