m-Tolyl isocyanate, 99%
m-Tolyl isocyanate, 99%
m-Tolyl isocyanate, 99%
Thermo Scientific Chemicals

m-Tolyl isocyanate, 99%

CAS: 621-29-4 | C8H7NO | 133.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L15255.06
also known as L15255-06
Price (USD)
32.65
Online Exclusive
35.90
Save 3.25 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
32.65
Online Exclusive
35.90
Save 3.25 (9%)
Each
Chemical Identifiers
CAS621-29-4
IUPAC Name1-isocyanato-3-methylbenzene
Molecular FormulaC8H7NO
InChI KeyCPPGZWWUPFWALU-UHFFFAOYSA-N
SMILESCC1=CC=CC(=C1)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5295-1.5320 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)>98.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only